Finnpipette® F2 GLP Kits, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Thermo Scientific Finnpipette® F2 GLP Kits provide durable, high-performance Finnpipette® F2 pipetters in an all-inclusive kit containing everything needed for pipetting.
The Thermo Scientific Finnpipette F2 Good Laboratory Pipetting kits offer the maximum reliability and comfort of the new Finnpipette F2. The F2 pipetters have unique features such as AVG (Advanced Volume Gearing mechanism) and a very large display for improved usability and comfort.
The Thermo Scientific Finnpipette® F2 Good Laboratory Pipetting kits offer the maximum reliability and comfort of the new Finnpipette® F2. The F2 pipetters have unique features such as AVG (Advanced Volume Gearing mechanism) and a very large display for improved usability and comfort.
Ordering information: In addition to the range of Finnpipette® F2 pipettors listed below, each kit includes F-Stand, F2 Pen, F2 Brochure, reagent reservoir demo pack, Good laboratory pipetting guide, and an assortment of compatible Finntip flex tips. Finntip flextips are the most advanced traditional tips, offering very low attachment and ejection forces. The Good laboratory pipetting guide contains essential pipetting information, from choosing the right type to detailed decontamination information.
Customers Who Bought This Also Bought