A propos de cet article
Thermo Scientific™ Finnpipette™ F1 GLP Kits offer a variety of single channels in a range of volumes that are packaged with compatible Finntips, and a convenient pipette stand to support good laboratory pipetting in your lab.
All new Thermo Scientific Finnpipette F1 Good Laboratory Pipetting kits offer the superior performance of the new Finnpipette F1. The F1 pipetters have unique features such as adjustable finger rest and antimicrobial surface for user comfort.
- Variety of single channels
- Range of volumes
- Packaged with compatible tips and stand
- Lightweight design
- Silent volume adjustment
- Secure volume lock
- Five-year warranty
Select the kit that is right for you, and take advantage of the Finnpipette F1 performance and lightweight design to maximize your comfort. Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI.
Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models. The secure volume setting prevents undesirable volume drift during operation.
Simply lift the plunger up to set volume and press down to lock in place with minimum effort. The 120° adjustable finger rest, for both right- and left-handed operator comfort, allows the hand to relax during pipetting cycles.
A 150% increase in air boost ensures efficient delivery of micro-volumes and prevent capillary action in 50 μl models and below.