Nalgene® Sanitary Gaskets for Nalgene Carboys; Silicon, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Sanitary gaskets for Nalgene carboys are recommended for use with 2630 and 2640 carboys series.
These gaskets can be used with closed-dome bio tanks, sanitary conical tanks, and bioprocess vessels. Autoclavable.
Protect against leakage, breakage and contamination by using Nalgene™ Silicone Sanitary Gaskets for Nalgene™ Carboys. Constructed of platinum-cured silicone, the gaskets ensure the highest degree of purity for biopharmaceutical applications.
Frequently Bought Together