Visitor Spec Center, NMC
# de catalogue 94003-854
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- Dispenser type:Bench top/wall-mountable
- No. of compartments:1
- PPE type:Safety glasses
- Description:Deluxe visitor specs dispenser
- Material:Acrylic
- Cat. no.:94003-854
- W×D×H:46×20×10 cm (18×8×4")
- Supplier No.:ASG-D
Spécifications
A propos de cet article
Acrylic dispenser features bottom door to keep glasses in place.
- Holds approximately four pairs
- Table or wall mount
Customers Who Bought This Also Bought