Acrylic Label Dispenser, NMC
Catalog # 94003-792
Supplier: ACCUFORM MANUFACTURING, INC
Specifications
- Description:Label dispenser, acrylic
- Cat. no.:94003-792
- Supplier No.:ALD1
Specifications
About this item
Organize and dispense your labels with ease
Ideal for labels on a roll diameter size up to a 4" width with a 3" core.
Customers Who Bought This Also Bought