Compact Glasses Holder, NMC
Catalog # 94003-848
Supplier: ACCUFORM MANUFACTURING, INC
Specifications
- Dispenser type:Bench top
- No. of compartments:1
- PPE type:Safety glasses
- Description:Compact dispenser for safety glasses without cover
- Cat. no.:94003-848
- W×D×H:7.62×22.86×15.24 cm (3×9×6")
- Supplier No.:ASG-1
Specifications
About this item
Designed to hold 4 - 6 pairs of glasses.
Customers Who Bought This Also Bought