Precoiled Pipe Markers 'Domestic Cold Water Supply', White/Green, National Marker
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
A propos de cet article
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas
Certifications: Meets ANSI A13.1-2007
Customers Who Bought This Also Bought