NMC Safety Tag Center
# de catalogue 94021-976
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- Description:Safety Tag Center
- Cat. no.:94021-976
- Dimensions:60.4 x 50.8 cm (24 x 20")
- Supplier No.:STC
Spécifications
A propos de cet article
Provide a convenient location to store your lockout tags
Made from high impact styrene. Features 43/4" deep tag pockets. Holds up to 800 safety tags.
Customers Who Bought This Also Bought