NMC Lockout Stations
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
A propos de cet article
High visibility stations facilitate proper and convenient lockouts
Heavy duty plexiglass board features replaceable hooks for locks and clear pockets for tags. Features lightweight Xenoy® lock bodies. Available in 48.26 x 60.96 cm (19 x 24") and 20.32 x 30.48 cm (8 x 12").
Customers Who Bought This Also Bought