Instatherm® Six-Foot Controller Cords, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Controller cords with various connectors for connecting all Ace Instatherm® vessels to temperature controllers.
Ordering information: Two versions (89075-444 and 71000-042) have "fast-acting" fuse to protect both the vessel and the controller from over-voltage or surges.
Customers Who Bought This Also Bought