Dispenser for Wipes and Bottles, S-Curve Technologies
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
These dispensers are designed to hold a supply of wipers and two bottles.
- Multiple choices to meet specific needs
- Wall mountable or free-standing
- Available without lid (wall-mountable option) or with clear wiper lid that protects wipers from contamination
- Flame polished edges for smoothness
Clear 1/4" PETG material.
Customers Who Bought This Also Bought