Finntip Stepper Tips, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Finntip stepper tips are the perfect fit with Thermo Scientific Finnpipette stepper pipetters.
The finntip stepper tips are plunger-style tips for positive displacement pipetting. Seven sizes are available in sterile and non sterile versions.
Customers Who Bought This Also Bought