Finntip™ Multistepper Pipette Tips, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Finntip Multistepper Pipette Tips optimize the performance of Thermo Scientific Finnpipette multistepper pipetters.
- Accommodates standard 9 mm microplate spacing
- Sterile or non-sterile versions
They are specifically designed for use with the Thermo Scientific Finnpipette Multistepper Pipetter. This tip has a nominal volume of 1500 μl and accommodates standard 9 mm microplate spacing. Finntip multistepper tips are available in racks of 96 tips, in both sterile and non-sterile versions.
Customers Who Bought This Also Bought