VWR® Vacuum Oven, Stainless Steel
Fournisseur: VWR International
Certificats
A propos de cet article
This vacuum oven is built with a stainless steel chamber for exceptional durability.
- PID digital temperature controller
- Digital vacuum switching gauge
- KF25 port as standard
- Independent OTL temperature limiter
VWR's double plenum design is energy efficient and promotes great temperature uniformity.
The doors on these units have positive latch handles with spring-loaded glass to facilitate a good vacuum seal without hinge binds that shorten the gasket life. A selection of gaskets (for specific applications) and a small benchtop footprint increase the versatility of these ovens.
The unique cross-flow ventilation forces inert gas to fill the entire chamber. To achieve required vacuum levels, users can choose from a 3/8" orifice or the KF25 fitting to withstand heavy use and minimize draw-down time. Maximum permitted end vacuum is 10 μ. Leak rate is 10 μ per hour.
Spécifications
- Catalog No:
- 97027-664
- 97027-674
- Temperature control:
- 0.1°C (32.18°F)
- 0.1°C (32.18°F)
- Temperature range:
- Ambient +10°C to 220°C (50 to 428°F)
- Ambient +10°C to 220°C (50 to 428°F)
- Temperature uniformity:
- ±6 % of Set point
- ±6 % of Set point
- Heat-up time:
- 90 min. to 150°C (302°F)
- 90 min. to 150°C (302°F)