Safety-Coated Glass Jugs, Clear
Fournisseur: Qorpak
Certificats
A propos de cet article
Safety coated glass jugs prevent lab mishaps, save time and reduce product liability.
- Ideal for handling and containing volatile substances
- Plastic-coating makes glass jugs easier to grip and harder to break
- If a safety coated jug breaks, the plastisol coating contains the substance in the jug long enough for proper disposal
Get the chemical resistance of glass with the safety of plastic.
Customers Who Bought This Also Bought