Envoy® Diagnostic Ready Engineered Shippers, Sonoco ThermoSafe
Fournisseur: Sonoco Thermosafe
Certificats
A propos de cet article
Envoy Diagnostic is a 2 to 8°C insulated shipping system that has been design-tested to ISCsilver™ ambient temperature profile to address the need for temperature assurance in moderate shipping conditions.
- Convenient all-in-one solution with proven thermal performance
- Ideal for 1- to 2-day shipments and small payloads
- Packouts incorporate foam bricks for uniform temperature coverage
Envoy Diagnostic is a 2 to 8°C insulated shipping system that has been design-tested to ISCsilver™ ambient temperature profile to address the need for temperature assurance in moderate shipping conditions.
A simple, economical solution for refrigerated temperature control of diagnostic kits, blood samples, and other diagnostic specimens.
Customers Who Bought This Also Bought