A propos de cet article
Nomadic® Pre-Qualified Shippers deliver precise protection for refrigerated and frozen pharmaceuticals, biological specimens, and samples in a range of configurations.
- Maintain 2–8°C for a minimum of 48 hours
- Maintain below -20°C for a minimum of 72 hours
- Replaceable components available
Nomadic® Pre-Qualified Shippers deliver precise protection for refrigerated and frozen pharmaceuticals, biological specimens, and samples in a range of configurations.
Nomadic® Pre-Qualified Shippers, 2–8°C shipping systems, have passed comprehensive thermal and transit testing performed by ISC Labs, the leading testing and validation laboratory in the industry. Shippers maintain minimum and maximum loads of refrigerated products at 2–8°C for a minimum of 48 hours, and below -20°C for a minimum of 72 hours with the use of dry ice pellets. Durable one-piece closed cell polyurethane foam insulated shipper
CFC and HCFC-free: Complies with international clean air regulations.