NS4 Series 1/4" Flow Dry Break, Quick Disconnect Coupling, Colder Products Company
Fournisseur: Avantor
Certificats
A propos de cet article
1/4" flow quick disconnect couplings offer high flow capacity in a compact size
Materials of construction include glass filled polypropylene, 316 stainless steel and EPDM oring seals.
Engineered with an advanced dry break non spill valve design to eliminate drips and mess enhancing operator and equipment safety. Glass filled polypropylebne for added strength under pressure and color coded for visual identfication.
Ordering information: Accessories not included
Customers Who Bought This Also Bought