Sanitary Fitting to Hose Barb Fitting Adapter, Colder Products Company
Fournisseur: Avantor
Certificats
A propos de cet article
Stainless steel adapter engineered to ease the transition from stainless steel process pipe and equipment to single use disposable plastic.
Materials of construction include 316L stainless steel and platinum cured, USP class 6 silicone oring seals.
Certifications: USP Class VI, BSE/TSE free materials (ADCF)
Ordering information: Accessories not included
Customers Who Bought This Also Bought