PMC Series 1/8" Flow Size, Quick Disconnect Coupling, Colder Products Company
Fournisseur: Colder Products
Certificats
A propos de cet article
1/8" flow quick disconnect couplings available in either polypropelene or acetal offer good chemical resistance for most applications
Materials of construction include polypropylene or acetal, 316 stainless steel and Buna N or EPDM oring seals.
Availabe with NSF listed materials for design flexibility.
Ordering information: Accessories not included
Customers Who Bought This Also Bought