CQG ChemQuik Series 3/8" Flow, Quick Disconnect Coupling, Colder Products Company
Fournisseur: Colder Products
Certificats
A propos de cet article
These 3/8" pressure balanced, non-spill Quick Disconnect Couplings are intended for high purity and aggressive chemical applications
Materials of construction include natural polypropylene, non-wetted Hastelloy C spring, FKM and FFKM o-ring seal options.
A non-metal flow path allows for a braod range of chemical compatibilities.
Ordering information: Accessories not included
Customers Who Bought This Also Bought