Quick Disconnect Coupling, APC Series ¼" Flow, Colder Products Company
Fournisseur: Avantor
Certificats
A propos de cet article
The 1/4" Flow APC series coupling features a plastic thumb latch, fewer moving parts and a smooth contoured design to deliver excelent flow in a compact size
Materials of construction include Acetal, 316 and 302 stainless steel, Buna-N o-ring seal.
The shrouded thumb latch protects against accidental disconnect.
Ordering information: Accessories not included
Customers Who Bought This Also Bought