VWR® Heavy-Duty Vacuum Bottles with Versatile Caps
Supplier: VWR International
Certificates
About this item
Heavy-duty vacuum bottles are designed to combine ease-of-use and versatility with the highest level of strength and durability.
- Excellent replacements for glass HPLC containers
- Available in HDPE and polypropylene formats
- Compatible with 50mm and 80mm versatile caps and adapters
Bottles feature thick walls that maintain full vacuum for 24 h, making them excellent replacements for glass HPLC containers, and ideal for applications involving liquid aspiration, cell harvesting, and degasification. They are compatible with 50mm and 80mm versatile caps and adapters, making it easy to customize the bottles to specific applications. Bottle material and volume are molded into the side of the bottle for easy identification, along with metric graduation marks certified accurate to ±5%. Polypropylene bottles, as well as all caps and adapters, are autoclavable at 15 PSI and 121°C for 15 min.
Customers Who Bought This Also Bought