A propos de cet article
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry. Each is manufactured to achieve the highest level of accuracy and precision possible.
These syringes are designed to fit into or with various different instruments such as the Microlab® 500 and Microlab® 1000.
Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze.
This series of syringes is ideal for dispensing volumes from 25μL up to 10mL. A variety of different terminations are available.
Gastight® syringes are ideal for dispensing both liquids and gases
Gastight® syringes are ideal for dispensing both liquids and gases
These syringes are designed to fit into or with various different instruments such as the Microlab® 500 and Microlab® 1000.
Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze.
This series of syringes is ideal for dispensing volumes from 25μL up to 10mL. A variety of different terminations are available.
This series of syringes is ideal for dispensing volumes from 25μL up to 10mL. A variety of different terminations are available.
Syringe Terminations
AccuDil (AD) - This termination is designed for Microlab® 1000 Diluters/Dispensers.
Cheminert (C or CX) - This termination is a 1/4-28 fitting designed to fit directly into instrumentation. The X-style, meaning plunger stop, on this syringe is designed to limit wear and tear on the plunger tip.
Diluter AccuDil (DAD) - This termination is designed for Microlab® 1000 Diluters/Dispensers and features a side port connector.
Diluter (DX) - This termination is designed for Microlab® 500 Diluters/Dispensers and features a side port connector.
PFTE Luer Lock with stop (TLLX) - Male luer taper with a nickel plated brass hub. This connects onto Microlab® 500 instruments.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 72310-396
- 25229-210
- 25229-212
- 72310-398
- 25229-214
- 60376-290
- 25229-216
- 25229-218
- 72310-020
- 25229-410
- 72310-018
- 25229-220
- 25229-222
- 72310-024
- 72310-400
- 72310-022
- 76437-220
- 25229-078
- 25229-136
- 89221-378
- 25229-103
- 25229-170
- 25229-012
- 25229-206
- 89221-384
- 25229-442
- 25229-008
- 25229-045
- 89221-466
- 60376-000
- 60376-004
- 60376-006
- 25229-351
- 25229-384
- 25229-412
- 60376-008
- 60376-018
- Type:
- Cheminert Syringes
- Cheminert Syringes
- Cheminert Syringes
- Cheminert Syringes
- Cheminert Syringes
- PFTE Luer Lock with Stop Syringes
- Cheminert Syringes
- Cheminert Syringes
- AccuDil Syringes
- PFTE Luer Lock with Stop Syringes
- AccuDil Syringes
- Cheminert Syringes
- Cheminert Syringes
- Diluter AccuDil Syringes
- Cheminert Syringes
- PFTE Luer Lock with Gastight Syringes
- Diluter Syringes
- Diluter Syringes
- Diluter Syringes
- Diluter Syringes
- Diluter Syringes
- Cheminert Syringes
- AccuDil Syringes
- PFTE Luer Lock with Stop Syringes
- PFTE Luer Lock with Stop Syringes
- Diluter Syringes
- Cheminert Syringes
- AccuDil Syringes
- AccuDil Syringes
- AccuDil Syringes
- PFTE Luer Lock with Stop Syringes
- PFTE Luer Lock with Stop Syringes
- Cheminert Syringes
- AccuDil Syringes
- AccuDil Syringes
- Autoclavable:
- Yes
- Yes
- Yes
- Yes
- Yes
- No
- Yes
- Yes
- No
- No
- No
- Yes
- Yes
- Yes
- Yes
- No
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- No
- No
- No
- Yes
- Yes
- No
- No
- No
- No
- No
- Yes
- No
- No
- Fluid path:
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE, PCTFE
- SST, Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- SST, Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- SST, Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE, PCTFE
- Borosilicate Glass, PTFE
- SST, Borosilicate Glass, PTFE
- SST, Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE, PCTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- SST, Borosilicate Glass, PTFE
- SST, Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Borosilicate Glass, PTFE
- Gas sterilisable:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. temperature:
- 115°C/239°F
- 115°C/239°F
- 115°C/239°F
- 115°C/239°F
- 115°C/239°F
- 115°C/239°F
- 115°C/239°F
- 115°C/239°F
- 50°C/122°F
- 115°C/239°F
- 50°C/122°F
- 115°C/239°F
- 115°C/239°F
- 50°C/122°F
- 115°C/239°F
- 80°C/176°F
- 115°C/239°F
- 115°C/239°F
- 115°C/239°F
- 115°C/239°F
- 115°C/239°F
- 115°C/239°F
- 50°C/122°F
- 115°C/239°F
- 115°C/239°F
- 115°C/239°F
- 115°C/239°F
- 50°C/122°F
- 50°C/122°F
- 50°C/122°F
- 115°C/239°F
- 115°C/239°F
- 115°C/239°F
- 50°C/122°F
- 50°C/122°F
- Min. temperature:
- 10°C/50°F
- 10°C/50°F
- 10°C/50°F
- 10°C/50°F
- 10°C/50°F
- 10°C/50°F
- 10°C/50°F
- 10°C/50°F
- 20°C/68°F
- 10°C/50°F
- 20°C/68°F
- 10°C/50°F
- 10°C/50°F
- 20°C/68°F
- 10°C/50°F
- 10°C/50°F
- 10°C/50°F
- 10°C/50°F
- 10°C/50°F
- 10°C/50°F
- 10°C/50°F
- 10°C/50°F
- 20°C/68°F
- 10°C/50°F
- 10°C/50°F
- 10°C/50°F
- 10°C/50°F
- 20°C/68°F
- 20°C/68°F
- 20°C/68°F
- 10°C/50°F
- 10°C/50°F
- 10°C/50°F
- 20°C/68°F
- 20°C/68°F