Seal Crimper, Labconco®
Fournisseur: Labconco
Certificats
A propos de cet article
Seal Crimper, seals Aluminum Seals on Serum Bottles with 13 mm or 20 corkage, as indicated
For use with FreeZone Freeze Dry Systems.
Customers Who Bought This Also Bought