VWR® Sealing Film, Polyester
Fournisseur: VWR International
Certificats
A propos de cet article
Polyester film, acrylic adhesive. These 49.5 µm thick polyester films minimise evaporation and prevent spillage and contamination between wells.
- Recommended temperature range from −40 to +120 °C
- Available sterile and non sterile
- Marginally pierceable by single channel pipette
- Applications: Short-term storage, incubation, ELISA
Each film L×D: 141,0×79,4 mm
Customers Who Bought This Also Bought