Spécifications
- Description:ScrubberMate™ Glassware and Rack Cart
- Cat. no.:97054-348
- Supplier No.:8027000
Spécifications
A propos de cet article
Designed for Labconco glassware washers, the ScrubberMate provides convenient storage and transport of extra Labconco washer racks, inserts or miscellaneous glassware
Rails are provided to allow a lower washer rack to slide directly onto the cart from the washer. In addition, the the lower drawer pulls out for simple loading of inserts or individual glassware. An upper washer rack may be docked above the upper drawer. The upper drawer accommodates two wire inserts and is removable so that tall glassware may be loaded in the lower drawer. Durable and chemical-resistant, high density polyethylene construction. Includes 2.5" diameter casters with toe locks.
Accommodates up to two Labconco washer racks or four wire inserts. Alternatively, 48 adjustable and removable pins, 0.5" wide x 4.75" high (1.3 cm x 12.1 cm), may be arrange in the drawers to hold inverted flasks, beakers and other glassware.