Hooded Combination Kjeldahl Digestion/Distillation Units, Labconco
Fournisseur: Labconco
Certificats
A propos de cet article
Hooded Combination Kjeldahl Digestion/Distillation Apparatus
Combines macro digestion and distillation processes for nitrogen determination of samples in one space-saving unit. Hood surrounding the apparatus helps contain the processing heat and vent it from the laboratory. Able to digest and distill up to six, twelve, or eighteen samples at one time. Hood is constructed of epoxy-coated formed steel with horizontal-sliding, tempered safety glass sashes. Frame is constructed of powder-coated tubular steel. Remote blowers are not included.
Ordering information: Requires remote exhaust blower, ductwork, and 500 or 800ml Kjeldahl flasks (not included). A remote auxiliary-air blower is recommended (not included). Units with 6, 12 and 18 places are available.
Customers Who Bought This Also Bought