Nalgene® Closures for Dropper Bottles, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Nalgene closures for dropper bottles fit 312750 and 312751 bottles and must be ordered with dispensing tip to complete Dropper Bottle System.
The closures are available in natural, white, yellow, green, red, and blue. Neck finish for 46300-568 and 46300-570 is 15-415.
Customers Who Bought This Also Bought