Nalgene® White Dropper Bottles, LDPE, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Nalgene White Dropper bottles are made of low-density polyethylene for reliable and repeatable dispensing.
The flexible dropper bottles offer excellent chemical resistance. The dropper control tip on the bottle snaps into place for a secure fit. The bottles are to be used with Nalgene® controlled dropper tips and closures (sold separately).
Customers Who Bought This Also Bought