Nalgene® Contact Clear Dropper Bottles, LDPE, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Nalgene Contact Clear dropper bottles are made of low-density polyethylene for reliable and repeatable dispensing.
These flexible dropper bottles offers excellent chemical resistance. The dropper control tip on the bottle snaps into place for a secure fit. The bottles are to be used with Nalgene® controlled dropper tips and closures (sold separately).
Customers Who Bought This Also Bought