ContactUsLinkComponent
Gastight® VDRL Antigen Dispenser Syringes, Hamilton
Gastight® VDRL Antigen Dispenser Syringes, Hamilton
# de catalogue 60384-104
Fournisseur:  Hamilton
Qualitative Syringe for Antigen Dispenser
Gastight® VDRL Antigen Dispenser Syringes, Hamilton
# de catalogue 60384-104
Fournisseur:  Hamilton
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Volume:
    833 µl
  • Description:
    Model VDRL 1/60ml LT Syringe
  • Supplier No.:
    87800
Spécifications
A propos de cet article
VDRL Antigen Dispensers are used with the PB600-1 Repeating Dispenser (sold separately) to dispense up to 50 times without refilling
Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision-machined PTFE plunger tip which creates a leak-free seal. The fluid path includes SST, borosilicate glass, and PTFE.
With the luer tip termination, needles are removable and fit over a ground glass hub which is tapered in the shape of a male luer. The LT termination will accept most hypodermic needles but was designed specifically for use with Hamilton Kel-F needles. This termination increases the dead volume in the syringe, which may not be appropriate for some applications. Autoclavable to 115°C (239°F) when disassembled.
This qualitative syringe delivers 1/60mL per dispense. A special 90° (bent) syringe needle (sold separately) allows easy syringe reading and filling.
Ordering information: Dispensers are also available as a 1/60mL quantitative or 1/100mL antigen dispenser. For these dispensers, the PB600-1 Repeating Dispenser (required), the special 90° (bent) syringe needle, or other available syringes, needle points, or gauges, contact your Avantor representative.
Customers Who Bought This Also Bought