Nalgene® Sealing Cap Assemblies, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Nalgene Sealing cap Assemblies seal Nalgene Oak Ridge centrifuge tubes and bottles
Cat. No. DS3131 features shoulder support ring to prevent shifting, binding or collapse of bottle neck. Before autoclaving, set closure on top of the container without engaging the threads.
Recommended for use at maximum speeds (10,000 x g or greater) without leakage, even when full. Also, recommended for spinning hazardous materials compatible with centrifuge tube material.
Customers Who Bought This Also Bought