ContactUsLinkComponent
Eye Tissue Region Single Species Blot, G-Biosciences
Eye Tissue Region Single Species Blot, G-Biosciences
Fournisseur:  G-Biosciences
Certificats
Eye Tissue Region Single Species Blot, G-Biosciences
Fournisseur:  G-Biosciences
Certificats
Spécifications
  • Catalog No:
  • CA82023-218
  • CA82023-220
  • Blot lane order:
  • 1. Protein marker
    2. Rat Whole Eye Lysate
    3. Rat retina lysate
    4. Rat cornea lysate
    5. Rat aqueous humor lysate
  • 1. Protein marker
    2. Rabbit Whole Eye Lysate
    3. Rabbit retina lysate
    4. Rabbit cornea lysate
    5. Rabbit aqueous humor lysate