Nalgene Suspended Magnetic Stir Bars, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Nalgene Floating Magnetic Stir Bars are designed for constant low-speed stirring action in tissue culture applications
The stir bars are ideal for tissue culture applications. The bar is slightly raised off the bottom surface of the vessel to minimize grinding effect on cells
Customers Who Bought This Also Bought