White Polypropylene Screw Caps, Unlined, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
White Unlined Polypropylene Caps are known for good impact strength, cost effectiveness, and pliability.
- Autoclavable
- Suitable for dry products
When autoclaving, place cap loosely on top of jar.
Customers Who Bought This Also Bought