White Polypropylene Flip Top Cap, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Flip Top Caps are ideal on the workbench for dispensing small amounts of liquid
They are made of durable polypropylene and are unlined. Orifice is 3.0mm.
Customers Who Bought This Also Bought