Green Polypropylene Hole Caps, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Polypropylene hole caps are available pre-assembled with septa or without septa.
- Good impact strength
- Cost effective
- Pliability
- Autoclavable
When autoclaving, set cap loosely on top of container.
Customers Who Bought This Also Bought