Qorpak® Black Phenolic Screw Caps, Pulp/Aluminum Foil Liner
Fournisseur: Qorpak
Certificats
A propos de cet article
Pulp/Aluminum Foil lined caps are typically used for food applications as they will not cause odor or taste problems
They can be used for non-acid, non-alkaline products. Great for organic solvents, chrome cleaners, brake fluids and mineral oils.
Customers Who Bought This Also Bought