Paint Cans, Epoxy Lined, Triple Tite Lid, Metal, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Epoxy Lined Round Paint Can with Triple Tite Lid provide a non-reactive barrier between the container and the product/atmosphere; protecting the contents and preventing rust
These cans are ideal for oil and water-based paints, glue, adhesive and stucco.
Customers Who Bought This Also Bought