Nalgene® Stackable Drawer, All-Purpose Acrylic, Thermo Scientific
# de catalogue 21020-236
Fournisseur: Thermo Fisher Scientific
Spécifications
- Description:Stackable drawer
- Cat. no.:21020-236
- Supplier No.:5832-0001
Spécifications
A propos de cet article
Nalgene® stackable drawers nest for high-density storage and can be mounted under a shelf or cabinet with provided screws.
- Clear acrylic organizers allows to view their contents easily
- Can withstand brief exposure to dilute acids, and bases
Helps to keep small lab items neatly organized and within reach.
Caution: Alcohols, organic solvents and strong acids may cause immediate damage.
Customers Who Bought This Also Bought