Packer Bottles, PET, Amber, Wide Mouth, Qorpak
Fournisseur: Qorpak
Certificats
A propos de cet article
PET Packer Bottles feature glass like clarity and are durable and rigid.
- PET packers are stress and impact resistant
- PET bottles are ideal for oil analysis
They have exceptional gas and moisture barrier properties and a good alcohol and solvent barrier.
Customers Who Bought This Also Bought