Jar, Polypropylene, White, Wide Mouth, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
White polypropylene jars are ideal for solids, powders and cosmetics.
Jars are ideal for storing and collecting specimens and lab supplies. Wide neck and straight sides facilitate easy recovery of contents. Polypropylene offers greater tolerance to high temperatures than HDPE and better contact clarity.
Customers Who Bought This Also Bought