Specifications
- Size:4 L (1 gal.)
- Cat. no.:28196-320
- Supplier No.:900-4000
Specifications
About this item
Nalgene L900 Liquid Detergent is formulated to thoroughly clean all plastic labware, especially polycarbonate
The detergent will not cause crazing, stress cracking or clouding when used as directed. This liquid detergent are for hand or machine use, it is biodegradable, not toxic, and low foaming.