Lab Style Bottles, High-Density Polyethylene, Narrow Mouth, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Narrow mouth HDPE lab style bottles are supplied with linerless polypropylene caps.
Excellent chemical resistance and the leak resistant closure make these ideal for storing, shipping, and packaging liquids for long periods. HDPE bottles are ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping. Linerless white polypropylene caps offer excellent impact strength, cost effectiveness, and pliability. Caps attached to the convenience pack and enclosed on the bulk pack
Customers Who Bought This Also Bought