Nalgene® 65 Braided Platinum-Cured Silicone Tubing, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Tubing is durable, tasteless, odorless, non-cytotoxic, non-pyrogenic, and non-hemolytic.
- Autoclavable
- Also be sterilized by gamma radiation
This high-purity tubing is designed for a variety of pump and transfer applications including pharmaceutical, laboratory, bioprocess manufacturing and food beverage applications.
Customers Who Bought This Also Bought