ContactUsLinkComponent
Handled Square Jugs, High-Density Polyethylene, Qorpak®
Handled Square Jugs, High-Density Polyethylene, Qorpak®
# de catalogue 76360-460
Fournisseur:  Qorpak
Jug Handled Square Hdpe 142 X 283Mm CS4
Handled Square Jugs, High-Density Polyethylene, Qorpak®
# de catalogue 76360-460
Fournisseur:  Qorpak
Spécifications
  • Capacity:
    3840 ml (128 oz.)
  • Dimensions:
    142×283 mm
  • Color:
    Natural
  • Description:
    White polypropylene F217 lined
  • Material:
    HDPE
  • Cat. no.:
    76360-460
  • Thread:
    38 - 400
Spécifications
A propos de cet article
Square jugs feature durable HDPE.
  • Excellent moisture barrier and are impact resistant
The space saving design on these jugs make them ideal for the workbench.
Customers Who Bought This Also Bought