Wide Mouth Rounds, High-Density Polyethylene
Fournisseur: Qorpak
Certificats
A propos de cet article
Natural HDPE wide mouth round bottles are ideal for general lab use with solids and powders.
- Available in multiple sizes, from 1 to 128 oz.
- Available with a variety of caps or no caps
- Bulk or convenience packs offered
These HDPE bottles provide an excellent moisture barrier and are impact resistant. They are ideal for high-speed filling, demanding pharmaceutical specifications, and the rough handling of interstate shipping.
Ordering information: Bottles 36319-300, 36319-302, 36319-359, 16125-812, 16125-820, and 93001-750 are supplied pre-cleaned for volatiles. Bottles 36319-342 and 76487-178 are supplied pre-cleaned and certified for volatiles. Bottles 76487-178, 76487-180, 36319-341, and 76539-382 are supplied pre-cleaned and certified for metals.
Customers Who Bought This Also Bought