Safety Coated Glass Jugs, Amber
Fournisseur: Qorpak
New Product
Certificats
A propos de cet article
Safety coated amber jugs with molded finger grips are the perfect storage solution for light sensitive products.
- Amber glass protects against harmful UV rays
- Jugs are slip and impact resistant, making them ideal for handling volatile substances
- Choose jugs only, or jugs with attached Black Phenolic F217 and PTFE lined caps
Customers Who Bought This Also Bought