Bottles, Cylinder, Natural High-Density Polyethylene, Narrow Mouth, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Natural HDPE commercial cylinder bottles are translucent and impact resistant.
They provide an excellent moisture barrier. Ideal for pharmaceutical applications. Narrow mouth cylinders are perfect for handling liquid samples in the lab. The shoulder style allows you to pour small quantities without spilling. HDPE bottles are ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping.
Ordering information: Bottles 36319-180 and 36319-182 are supplied pre-cleaned for volatiles. Bottle 36319-171 is supplied pre-cleaned and certified for volatiles. Bottles 93001-754, 93001-758 and 76487-176 are supplied pre-cleaned and certified for metals. Bottle TNPLC09178 is supplied with Vacuum and Ionization treatment.
Customers Who Bought This Also Bought